missing translation for 'onlineSavingsMsg'
Learn More

PP1 alpha/PPP1A Antibody (4G3), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18414555 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414555 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414555 Supplier Novus Biologicals Supplier No. NBP104334AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PP1 alpha/PPP1A Monoclonal antibody specifically detects PP1 alpha/PPP1A in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP1 alpha/PPP1A
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 4G3
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.16, PP1alpha, PP-1AMGC15877, PPP1AMGC1674, protein phosphatase 1, catalytic subunit, alpha isoform, protein phosphatase 1, catalytic subunit, alpha isozyme, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, serine/threonine-protein phosphatase PP1-alpha catalytic subunit
Host Species Mouse
Immunogen Recombinant human PP1 alpha/PPP1A (30-299aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5499
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.