missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PP1 alpha Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583841
This item is not returnable.
View return policy
Description
Immunogen sequence: QILKPADKNK GKYGQFSGLN PGGRPIT Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
This gene encodes a reticuloplasmin, a protein which resides in the lumen of the endoplasmic reticulum. The protein shows sequence similarity to the protein disulfide isomerase family. However, it lacks the thioredoxin motif characteristic of this family, suggesting that this protein does not function as a disulfide isomerase. The protein dimerizes and is thought to play a role in the processing of secretory proteins within the ER. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| PP1 alpha | |
| Polyclonal | |
| Unconjugated | |
| PPP1CA | |
| 96A; CG6593; CG6593-PA; CG6593-PB; dism2; Dmel\CG6593; Dmel_CG6593; DmPp1-96A; GADD34; LD14639p; MGC15877; MGC1674; PP1; PP1 96A; PP1 alpha96a; Pp1-96A; PP1A; PP-1A; PP1A-96A; PP1alpha; PP1alpha[[1]]; PP1alpha96A; pp1alpha-96A; Pp1alpha-96A-PA; Pp1alpha-96A-PB; PP1c; PP1c 96A; PPP1A; Ppp1c; Ppp1ca; protein phosphatase 1; protein phosphatase 1 catalytic subunit alpha; protein phosphatase 1, catalytic subunit, alpha isoform; protein phosphatase 1, catalytic subunit, alpha isozyme; Protein phosphatase 1alpha at 96A; protein phosphatase type 1 alpha catalytic subunit; Protein phosphatase type 1 alpha, catalytic subunit; protein phosphatase-1d; serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit; Serine/threonine-protein phosphatase alpha-1 isoform; Serine/threonine-protein phosphatase PP1-alpha catalytic subunit; testicular tissue protein Li 155 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5499 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P62136 | |
| PPP1CA | |
| Recombinant protein corresponding to Human PPP1CA. Recombinant protein control fragment (Product #RP-102999). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction