missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PP 2
GREENER_CHOICE

Product Code. 30001245
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30001245

Brand: Tocris Bioscience™ 1407/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective Src family kinase inhibitor

Selective inhibitor of Src-family tyrosine kinases. Inhibits p56lck and p59fynT (IC50 values are 4 and 5 nM respectively). Displays > 10000-fold selectivity over ZAP-70 and JAK2. Moderately inhibits CSK (IC50 = 0.73 μM).

Negative Control also available.

TRUSTED_SUSTAINABILITY

Specifications

CAS 172889-27-9
Quantity 10 mg
Molecular Formula C15H16ClN5
Formula Weight 301.78
Purity >99%
Molecular Weight (g/mol) 301.778
Chemical Name or Material PP 2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.