missing translation for 'onlineSavingsMsg'
Learn More

PP 121
GREENER_CHOICE

Product Code. p-7116049
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18755856

Brand: Tocris Bioscience 3894/25

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Dual kinase inhibitor; inhibits PI 3-K family kinases

PP 121 is a dual inhibitor of receptor tyrosine kinases (RTKs) (IC50 < 0.02 μM for Abl, Src, VEGFR-2 and PDGFR) and PI 3-K family kinases (IC50 < 0.06 μM for p110α, DNA-PK and mTOR). Exhibits no significant effect on receptor serine/threonine kinases (RSTKs). Blocks the proliferation of tumor cells by direct inhibition of PI 3-K, mTOR, Src and the VEGF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Cyclopentyl-3-(1H-pyrrolo[2,3-b]pyridin-5-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
CAS 1092788-83-4
Assay Percent Range >99%
Quantity 25 mg
Molecular Formula C17H17N7
Formula Weight Observed MW: 319.36
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.