missing translation for 'onlineSavingsMsg'
Learn More

PP 121

Product Code. 18755856
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18755856 25 mg 25mg
18749206 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18755856 Supplier Tocris Bioscience Supplier No. 3894/25

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dual kinase inhibitor; inhibits PI 3-K family kinases

PP 121 is a dual inhibitor of receptor tyrosine kinases (RTKs) (IC50 < 0.02 μM for Abl, Src, VEGFR-2 and PDGFR) and PI 3-K family kinases (IC50 < 0.06 μM for p110α, DNA-PK and mTOR). Exhibits no significant effect on receptor serine/threonine kinases (RSTKs). Blocks the proliferation of tumor cells by direct inhibition of PI 3-K, mTOR, Src and the VEGF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Cyclopentyl-3-(1H-pyrrolo[2,3-b]pyridin-5-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
CAS 1092788-83-4
Assay Percent Range >99%
Quantity 25 mg
Product Line Tocris
Molecular Formula C17H17N7
Formula Weight Observed MW: 319.36
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.