missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Poxygrid Test Tube Rack

Product Code. 17715834 Shop All SP Scienceware Products
Click to view available options
Positions:
48
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17715834

Brand: Bel Art™ F187840001

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy. Orange racks can be used for biohazard warning, yellow for radioactive warning

  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F); autoclavable
  • All racks have square holes, three decks, a slotted grid bottom and reinforced edges

Specifications

Product Type Test Tube Rack
Array 6 x 8
Autoclavable Autoclavable
Color Blue
Holds Test Tubes
Material Steel Wire
Positions 48
Shape Rectangular
Shelves 3 Tiers
Stackable Yes, When Empty
Sterility Non-sterile
No. per Pack 1 Ea.
Dimensions (L x W x H) 36.5 x 30.4 x 10.2 cm
Height (Metric) 10.2 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.