missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Poxygrid Steel Test Tube Racks

Chemical and corrosion resistant steel test tube racks in different colors for easy identification.

Brand:  Bel-Art™ SP Scienceware™ F18754-0001

38.86 GBP valid until 2024-04-26
Use promo code "21699" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.19050kg

 View more versions of this product

Product Code. 15580012

  • £40.90 / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

Chemical and corrosion resistant steel test tube racks in different colors for easy identification.

  • Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy
  • Orange racks can be used for biohazard warning, yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of −223°C (−369°F) to 121°C (250°F)
  • All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications

Specifications

Test Tube Rack
Autoclavable
Tubes (13 to 16 mm)
40
Non-sterile
-223°C to +121°C
16 mm
4 x 10
Blue
Epoxy Coated Steel
Rectangle
13 - 16mm Tubes
20.3 x 9.2 x 6.4 cm (8 x 3.62 x 2.50 in.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers