missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Powerflow™ Face-Mounted Powered Air Respirator System
Click to view available options
:
Color: Black
Description
- Eye, face, and respiratory protection against hazards present when stripping and abating asbestos
- Side-mounted P3 filter for use against solid and liquid aerosols
- Compact, face-mounted 3M™ Powerflow™ Turbo Unit, PFT-01 with DIN knuckle threads
- Six strap harness for a secure fit and quick release buckles for ease of removal
- Soft, pliable silicone face mask for user comfort
- Integrated speech diaphragm to improve communication
- Large, polycarbonate lens for a wide field of view
- Lightweight lithium-ion rechargeable battery holds up to five hours of power and is IP54 rated
- Intelligent fast charging battery charger included
Specifications
Specifications
| Assigned Protection Factor (APF) | 1000 |
| Battery Type | Lithium-ion |
| Cartridge/Filter | PARTICULATE |
| Certifications (CSA) | CE Approved |
| Certifications (EN) | EN 136:1998 (Class 2), EN 166:2001, EN 14594 3B |
| Color | Black |
| Product Type | PAPR System with Facepiece |
| EN Approval EN12941 | Yes |
| Model | Powerflow™ |
| Mounting Type | Face-mounted |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction