Learn More
3M™ Powered Air Respirator System Starter Kit PF-619E
Click to view available options
:
PAPR System; Design Type: Belt-Mounted
Description
- Starter Kit includes a 3M™ PF-600E turbo unit, charger and belt.
- Compatible with the 3M™ DT Series filters and specific 3M™ Versaflo™ headtops
- Audible and visual alarms alert the user to low battery or if the flow rate falls below the manufacturer's minimum design flow rate
- Durable design suitable for cleaning by shower/water spray
- Integrated battery and simple one-button functionality
Specifications
Specifications
| Battery Life | 4 to 8 hours |
| Battery Type | Lithium-ion |
| Description | 3M™ Powered Air Respirator System Starter Kit PF-619E |
| Product Type | PAPR Assembly, No Headtop |
| Facepiece/Headtop Model | No Headgear Included |
| Intrinsic Safety (IS) Rating | No |
| Model | PF-619E |