missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ POWERCOAT™ Nitri-Knit Gloves
Description
- Supported nitrile glove
- Cotton interlock liner
- Textured finish
- Interlock knit liner gives gloves added strength for resistance to cuts, snags, abrasions and punctures while adding comfort to wearer
- Hand portion is treated with a permanent anti-bacterial and anti-mildew agent
- Unique textured finish provides excellent grip on oily, wet or dry objects
- High nitrile content offers superior resistance to cuts, snags, abrasions and punctures
- Outperforms supported natural rubber, neoprene and vinyl gloves when exposed to solvents, caustics and animal fats
- True dip coated glove has a permanent bond of nitrile to inner-liner, won't peel apart like glued-in liners
- Gloves are free of latex proteins which can cause allergic reactions
Specifications
Specifications
| Certifications/Compliance | EC PPE Category 3, EC Certificate/Attestation Number BP60077212 0001 |
| Coating Material | Nitrile |
| Color | Blue |
| Exterior Finish | Fully Textured |
| Size | 9, Large |
| Thickness (Metric) | 1 mm |
| Liner Material | Cotton |
| Length (Metric) | 30 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?