Learn More
Thermo Scientific™ Power Cords for myECL Imager and Pierce G2 Fast Blotter
These electrical power cords have a C/13 connector at one end and a specific plug configuration at the opposite end to match electrical outlets of specific countries.
Brand: Thermo Scientific™ 84859
Code : PN
Additional Details : Weight : 0.50000kg
Description
Thermo Scientific™ Power Cords for myECL Imager and Pierce G2 Fast Blotter have a C/13 connector at one end and a specific plug configuration at the opposite end to match electrical outlets of specific countries.
Additional Information and Online Ordering »
The black, 2.4- to 3.5m-long, cords have been validated to replace the cordset that is supplied with particular Thermo Scientific powered devices. Specifically, these instruments include the myECL Imager (Part No. 62236) and the Pierce G2 Fast Blotter (Part No. 62288).
Highlights:
- Black
- 2.4 to 3.5m long
- C13 connector
- Country-specific connectors
Specifications
Pierce Power System, myECL Imager | |
Power Cord with C/13 Connector, Continental Europe | |
2.4 to 3.5 m | |
Black | |
Replacement of myECL Imager and Pierce G2 Fast Blotter accessory |
Power Cord | |
94.48 to 137 in. | |
Black 2.5 m power cord | |
Protein biology, protein gel electrophoresis, protein assays and analysis, western blotting | |
1 Cord |