missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Power Blotter Pre-cut Membranes and Filters, nitrocellulose, regular size

Precut, preassembled nitrocellulose membrane and filter paper sandwiches for gels

Brand:  Invitrogen™ PB7320

Additional Details : Weight : 0.15200kg

 View more versions of this product

Product Code. 15858502

  • £253.00 / Pack of 20
In Stock
to see stock.
Add to basket

Includes: The pre-cut membranes and filters come with 20 sheets of nitrocellulose and 80 sheets of absorbant blotting paper (0.85mm thickness)

Description

Description

The Invitrogen Power Blotter Pre-cut Membranes and Filters are designed for transferring proteins from gels using the Invitrogen Power Blotter System.

Features:

  • Membranes and filters allow for high efficiency semi-dry transfer of proteins in less than 10 minutes when combined with the Power Blotter 1-Step Transfer Buffer
  • 100% pure nitrocellulose membrane (0.2 μM pore size) with high surface area and excellent uniformity provides high-quality transfers.
    • Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods such as chemiluminescence, chromogenic, and fluorescence. 
    • 209 μg/cm2 protein binding affinity
    • Blocks easily produces very low background in chemiluminescent and fluorescent western blotting
  • Convenient— Pre-cut sheets are ready to use, meaning no need to cut your own membranes, minimizing damage and contamination
  • Validated – When combined, membrane and filter provide the thickness and buffer capacity needed for optimal performance
Specifications

Specifications

13 cm
Transfer Stack
0.2 μm
Room Temperature
0.85 mm
20 Blots
8.5 cm
Nitrocellulose
20 regular-size transfer stacks (13 cm x 8.3 cm) composed of 20 sheets of nitrocellulose and 80 sheets of filter paper (0.85 mm thickness)
Regular, 13 cm x 8.5 cm
Power Blotter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.