Learn More
Invitrogen™ Power Blotter Pre-cut Membranes and Filters, nitrocellulose, mini
Precut, preassembled nitrocellulose membrane and filter paper sandwiches for gels
Brand: Invitrogen™ PB7220
Additional Details : Weight : 5.80000kg
Includes: The pre-cut membranes and filters come with 20 sheets of nitrocellulose and 80 sheets of absorbant blotting paper (0.85mm thickness)
Description
The Invitrogen Power Blotter Pre-cut Membranes and Filters are designed for transferring proteins from gels using the Invitrogen Power Blotter System.
Features:
- Membranes and filters allow for high efficiency semi-dry transfer of proteins in less than 10 minutes when combined with the Power Blotter 1-Step Transfer Buffer
- 100% pure nitrocellulose membrane (0.2 μM pore size) with high surface area and excellent uniformity provides high-quality transfers.
- Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods such as chemiluminescence, chromogenic, and fluorescence.
- 209 μg/cm2 protein binding affinity
- Blocks easily produces very low background in chemiluminescent and fluorescent western blotting
- Convenient— Pre-cut sheets are ready to use, meaning no need to cut your own membranes, minimizing damage and contamination
- Validated – When combined, membrane and filter provide the thickness and buffer capacity needed for optimal performance
Specifications
7 cm | |
Transfer Stack | |
0.45 μm | |
Room Temperature | |
0.85 mm | |
20 Blots |
8.4 cm | |
Nitrocellulose | |
20 mini-size transfer stacks (8 cm x 8 cm) composed of 20 sheets of nitrocellulose and 80 sheets of filter paper (0.85 mm thickness) | |
Mini, 7 cm × 8.4 cm | |
Power Blotter |
For Research Use Only. Not for use in diagnostic procedures.