missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gammex™ Powder-free Sterile Latex Surgical Gloves
Click to view available options
Size:
5.5
7
Unit Size:
Pack of 40
Description
- Sterile
- Powder-free
- Contains latex
- Highly resistant to tears, alcohol based hand disinfectants and viral penetration
- Very low protein content - thiuram and MBT-free
Specifications
Specifications
| Color | Natural |
| Latex Free | No |
| Powdered | Powder Free |
| Size | 5.5 |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction