missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gammex™ Powder-free Sterile Latex Surgical Gloves
Description
- Sterile
- Powder-free
- Contains latex
- Highly resistant to tears, alcohol based hand disinfectants and viral penetration
- Very low protein content - thiuram and MBT-free
Specifications
Specifications
| Color | Natural |
| Latex Free | No |
| Powdered | Powder Free |
| Size | 5.5 |
| Sterility | Sterile |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?