Learn More
Invitrogen™ Powassan Virus NS1 Monoclonal Antibody (3681)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547516
Description
Specificity: Specific for the NS1 of Powassan virus. Negative vs. related viruses. Nonreactive with: West Nile virus, Eastern Equine Encephalitis virus, Western Equine Encephalitis virus, St. Louis Encephalitis virus and La Crosse virus. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Powassan virus is a tickborne flavivirus.
Specifications
| Powassan Virus NS1 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Proprietary sequence. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 3681 | |
| Unconjugated | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.