Learn More
Lenz Laborglasintrumente Pouring Rings PBT Red
Product Code. 10798271
Click to view available options
Thread Size:
GL 32
GL 45
Unit Size:
Each
Product Code. 10798271
Brand: Lenz Laborglasintrumente 07105600
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Material: PBT (red)
  • Temperature resistance: up to 180 °C
Specifications
Color Red
Material PBT
Thread Size GL 45