missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Pouch Cap for Polymer Pouch

Product Code. 15829856 Shop All Applied Biosystems Products
Click to view available options
Quantity:
4 Pieces
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15829856

Brand: Applied Biosystems™ 4412619

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This is a cap for use with POP-4™, POP-6™, and POP-7™ polymer pouches.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each package contains 4 caps.
Format Pouch
For Use With (Equipment) 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
For Use With (Application) Sequencing
Product Type Pouch Cap for Polymer Pouch
Quantity 4 Pieces
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.