missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU6F1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38408-100ul
This item is not returnable.
View return policy
Description
POU6F1 Polyclonal antibody specifically detects POU6F1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| POU6F1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 | |
| Brain-specific homeobox/POU domain protein 5, Brn-5, BRN5brain-5, mPOU homeobox protein, MPOUbrn-5, POU class 6 homeobox 1, POU domain, class 6, transcription factor 1, TCFB1Brain-5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-301 of human POU6F1 (NP_002693.3).,, Sequence:, MPGISSQILTNAQGQVIGTLPWVVNSASVAAPAPAQSLQVQAVTPQLLLNAQGQVIATLASSPLPPPVAVRKPSTPESPAKSEVQPIQPTPTVPQPAVVIASPAPAAKPSASAPIPITCSETPTVSQLVSKPHTPSLDEDGINLEEIREFAKNFKIRRLSLGLTQTQVGQALTATEGPAYSQSAICRFEKLDITPKSAQKLKPVLEKWLNEAELRNQEGQQNLMEFVGGEPSKKRKRRTSFTPQAIEALNAYFEKNPLPTGQEITEIAKELNYDREVVRVWFCNRRQTLKNTSKLNVFQIP | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 5463 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction