missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU6F1 Antibody (6F10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00005463-M03
This item is not returnable.
View return policy
Description
POU6F1 Monoclonal antibody specifically detects POU6F1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| POU6F1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Brain-specific homeobox/POU domain protein 5, Brn-5, BRN5brain-5, mPOU homeobox protein, MPOUbrn-5, POU class 6 homeobox 1, POU domain, class 6, transcription factor 1, TCFB1Brain-5 | |
| POU6F1 (NP_002693, 193 a.a. ~ 301 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ITPKSAQKLKPVLEKWLNEAELRNQEGQQNLMEFVGGEPSKKRKRRTSFTPQAIEALNAYFEKNPLPTGQEITEIAKELNYDREVVRVWFCNRRQTLKNTSKLNVFQIP | |
| 0.1 mg | |
| DNA replication Transcription Translation and Splicing | |
| 5463 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 6F10 | |
| Western Blot 1:500, ELISA | |
| NP_002693 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction