missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POU5F1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566155
This item is not returnable.
View return policy
Description
Immunogen sequence: VGPGSEVWGIP PCPPPYELCG GMAYCGPQVG VGLVPQGGLE TSQPESEAGV Highest antigen sequence identity to the following orthologs - mouse 76%, rat 76%.
This intronless gene was thought to be a transcribed pseudogene of POU class 5 homeobox 1, however, it has been reported that this gene can encode a functional protein. The encoded protein is nearly the same length as and highly similar to the POU class 5 homeobox 1 transcription factor, has been shown to be a weak transcriptional activator and may play a role in carcinogenesis and eye development.
Specifications
| POU5F1B | |
| Polyclonal | |
| Unconjugated | |
| POU5F1B | |
| OCT4PG1; Oct4-pg1; octamer binding protein 3_like sequence; Octamer-binding protein 3-like; Octamer-binding transcription factor 3-like; OTF3C; OTF3P1; POU 5 domain protein; POU class 5 homeobox 1 pseudogene 1; POU class 5 homeobox 1B; POU domain transcription factor Oct-4; POU domain transcription factor OCT4-pg1; POU domain, class 5, transcription factor 1 pseudogene 1; POU5F1B; POU5F1P1; POU5F1P4; POU5FLC20; POU5FLC8; Putative POU domain, class 5, transcription factor 1B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5462 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q06416 | |
| POU5F1B | |
| Recombinant Human POU5F1B. Recombinant protein control fragment (Product #RP-105098). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction