Learn More
POU4F3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21509-1-AP-20UL
 View more versions of this product
Product Code. 16891095
  • £135.00 / 20µL
Description
This gene encodes a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. This protein is found in the retina and may play a role in determining or maintaining the identities of a small subset of visual system neurons. Defects in this gene are the cause of non-syndromic sensorineural deafness autosomal domit type 15.
Specifications
Specifications
POU4F3
Polyclonal
Unconjugated
POU4F3
Brain 3C, Brn 3C, BRN3C, DFNA15, POU class 4 homeobox 3, POU4F3
Rabbit
Antigen Affinity Chromatography
RUO
18998, 5459
-20°C
Liquid
Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15319, Q63955
Pou4f3
POU4F3 Fusion Protein Ag16004
20 μL
Primary
Mouse, Human