missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU4F3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38456-20ul
This item is not returnable.
View return policy
Description
POU4F3 Polyclonal antibody specifically detects POU4F3 in Human samples. It is validated for ELISA,Western Blot
Specifications
| POU4F3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| brain-3C, Brain-specific homeobox/POU domain protein 3C, BRN3.1, Brn-3C, BRN3Cbrn-3C, DFNA15, MGC138412, POU class 4 homeobox 3, POU domain class 4, transcription factor 3, POU domain, class 4, transcription factor 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human POU4F3 (NP_002691.1).,, Sequence:, MMAMNSKQPFGMHPVLQEPKFSSLHSGSEAMRRVCLPAPQLQGNIFGSFDESLLARAEALAAVDIVSHGKNHPFKPDATYHTMSSVPCTSTSSTVPISHPAALTSHPHHAVHQGLEGDLLEHISPTLSVSGLGAPEHSVMPAQIHPHHLGAMGHLHQAMGMSHPHTVAPHSAMPACLSDV | |
| 20 μL | |
| Neuroscience, Vision | |
| 5459 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction