missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU3F3 Polyclonal Antibody

POU3F3 Polyclonal Antibody
Brand: Invitrogen PA565740
This item is not returnable.
View return policy
Description
POU3F3 Polyclonal Antibody for ICC/IF
Specifications
| POU3F3 | |
| Polyclonal | |
| Unconjugated | |
| POU3F3 | |
| 8-Oct; Brain-1; brain-specific homeobox/POU domain protein 1; Brn1; Brn-1; brn-1 protein; class III POU protein POU-domain; oct-8; Octamer-binding protein 8; octamer-binding transcription factor 8; Otf8; OTF-8; POU class 3 homeobox 3; POU domain, class 3, transcription factor 3; Pou3f3; RHS1; Rhs2; Skin1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5455 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20264 | |
| POU3F3 | |
| Recombinant Human POU3F3. Recombinant protein control fragment (Product #RP-104002). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction