missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POU3F3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564311
This item is not returnable.
View return policy
Description
Immunogen sequence: GGAQSLVHPG LVRGDTPELA EHHHHH Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
POU3F3 is a member of the class III POU family of transcription factors that are expressed in the central nervous system. The POU domain in these proteins is required for high affinity binding to octamer DNA sequences.
Specifications
| POU3F3 | |
| Polyclonal | |
| Unconjugated | |
| POU3F3 | |
| 8-Oct; Brain-1; brain-specific homeobox/POU domain protein 1; Brn1; Brn-1; brn-1 protein; class III POU protein POU-domain; oct-8; Octamer-binding protein 8; octamer-binding transcription factor 8; Otf8; OTF-8; POU class 3 homeobox 3; POU domain, class 3, transcription factor 3; Pou3f3; RHS1; Rhs2; Skin1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5455 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20264 | |
| POU3F3 | |
| Recombinant protein fragment of Human POU3F3 (Product #RP-104002) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction