missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POTEH, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00023784-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MVAEAGSMPAASSVKKPFGLRSKMGKWCRHCFAWCRGSGKSNVGTSGDHDDSAMKTLRSKMGKWCCHCFPWCRGSGKSNVGTSGDHDDSAMKTLRSKMGKWCCHCFPCCRGSGKSNVGTSGDHDDSAMKTLRSKMGKWCCHCFPCCRGSGKNKVGPWGDYDDSAFMEPRYHVRREDLDKLHRAAWWGKVPRKDLIVMLKDTDMNKKDKQKRTALHLASANGNSEVVKLLLDRRCQLNILDNKKRTALTKAVQCQEDECALMLLEHGTDPNIPDEYGNTALHYAIYNEDSpecifications
| POTEH | |
| Polyclonal | |
| No additive | |
| BC148758 | |
| POTEH | |
| ACTBL1 (AAI48759.1, 1 a.a. ∼ 288 a.a) full-length human protein. | |
| RUO | |
| 23784 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| POTE ankyrin domain family, member H | |
| A26C3/ACTBL1/POTE22 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction