missing translation for 'onlineSavingsMsg'
Learn More

POTEG Antibody (OTI2G1), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-73540PCP

Product Code. 30032126

  • £561.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

POTEG Monoclonal antibody specifically detects POTEG in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

POTEG
Monoclonal
PerCP
PBS
Mouse
Immunogen affinity purified
RUO
404785
Store at 4C in the dark.
IgG1
Western Blot, Immunohistochemistry
OTI2G1
Western Blot, Immunohistochemistry
A26C2, ANKRD26-like family C, member 2, cancer/testis antigen family 104, member 4, CT104.4, POTE ankyrin domain family member G, POTE ankyrin domain family, member G, POTE14ACTBL1, POTE14alpha, POTE-14ANKRD26-like family C member 2, POTE22, Prostate, ovary, testis-expressed protein on chromosome 14, protein expressed in prostate, ovary, testis, and placenta 14
Full length human recombinant protein of human POTEG (NP_001005356) produced in HEK293T cell.
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.