missing translation for 'onlineSavingsMsg'
Learn More

POTEG Antibody (OTI2G1), PE, Novus Biologicals™

Product Code. 30024887 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30024887 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30024887 Supplier Novus Biologicals Supplier No. NBP273540PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

POTEG Monoclonal antibody specifically detects POTEG in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen POTEG
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2G1
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias A26C2, ANKRD26-like family C, member 2, cancer/testis antigen family 104, member 4, CT104.4, POTE ankyrin domain family member G, POTE ankyrin domain family, member G, POTE14ACTBL1, POTE14alpha, POTE-14ANKRD26-like family C member 2, POTE22, Prostate, ovary, testis-expressed protein on chromosome 14, protein expressed in prostate, ovary, testis, and placenta 14
Host Species Mouse
Immunogen Full length human recombinant protein of human POTEG (NP_001005356) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404785
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.