missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POTEG Antibody (OTI2G1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73540B
This item is not returnable.
View return policy
Description
POTEG Monoclonal specifically detects POTEG in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| POTEG | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| POTEG | |
| Full length recombinant protein of human POTEG (NP_001005356) produced in HEK293T cells. | |
| 0.1 mL | |
| 404785.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2G1 | |
| Western Blot, Immunohistochemistry | |
| A26C2, ANKRD26-like family C, member 2, cancer/testis antigen family 104, member 4, CT104.4, POTE ankyrin domain family member G, POTE ankyrin domain family, member G, POTE14ACTBL1, POTE14alpha, POTE-14ANKRD26-like family C member 2, POTE22, Prostate, ovary, testis-expressed protein on chromosome 14, protein expressed in prostate, ovary, testis, and placenta 14 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction