missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POTEE Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54706
This item is not returnable.
View return policy
Description
POTEE Polyclonal specifically detects POTEE in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| POTEE | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| A26C1, A26C1A, ANKRD26-Like Family C Member 1A, ANKRD26-Like Family C, Member 1A, Cancer/Testis Antigen Family 104, Member 2, CT104.2, POTE Ankyrin Domain Family Member E, POTE2, POTE-2, POTE2gamma, Prostate, Ovary, Testis-Expressed Protein On Chromosome 2, Protein Expressed In Prostate, Ovary, Testis, And Placenta 2 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 445582 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| POTEe | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:LKALQLNELTMDDDTAVLVIDNGS | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction