missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POTEB Polyclonal Antibody

Description
Antibody detects endogenous levels of total A26B1.
POTEB (POTE Ankyrin Domain Family Member B) is a Protein Coding gene. An important paralog of this gene is POTEB2.
Specifications
Specifications
| Antigen | POTEB |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | POTEB |
| Gene Accession No. | A0A0A6YYL3 |
| Gene Alias | A26B1; ANKRD26-like family B member 1; ANKRD26-like family B, member 1; cancer/testis antigen family 104, member 5; CT104.5; POTE ankyrin domain family member B; POTE ankyrin domain family member B2; POTE ankyrin domain family member B-like; POTE ankyrin domain family, member B; POTE15; POTE-15; POTEB; POTEB2; Prostate, ovary, testis-expressed protein on chromosome 15; protein expressed in prostate, ovary, testis, and placenta 15 |
| Gene Symbols | POTEB |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?