missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POTEA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20203-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to POTEA.
POTE8 (Prostate, ovary, testis-expressed protein on chromosome 8), also known as A26A1 (ANKRD26-like family A member 1) or POTEA (POTE ankyrin domain family, member A), is a 498 amino acid protein that contains five ANK repeats and is thought to participate in signaling events during reproductive development. POTE8 exists as two alternatively spliced isoforms which are encoded by a gene that maps to human chromosome 8. Consisting of nearly 146 million base pairs, chromosome 8 encodes over 800 genes and is associated with a variety of diseases and maligcies. Schizophrenia, bipolar disorder, Trisomy 8, Pfeiffer syndrome, congenital hypothyroidism, Waardenburg syndrome and some leukemias and lymphomas are thought to occur as a result of defects in specific genes that maps to chromosome 8.Specifications
| POTEA | |
| Polyclonal | |
| Unconjugated | |
| POTEA | |
| A26A1, ANKRD26 like family A member 1, POTE 8, POTE8, POTEA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 340441 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6S8J7 | |
| POTEA | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction