missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POTEA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539361
This item is not returnable.
View return policy
Description
Specifications
| POTEA | |
| Polyclonal | |
| Unconjugated | |
| POTEA | |
| POTEA | |
| A synthetic peptide derived from the C-terminal region of human POTE8 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6S8J7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 340441 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction