missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
POTEA Polyclonal antibody specifically detects POTEA in Human samples. It is validated for ELISA,Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | POTEA |
| Applications | ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Dilution | ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | A26A1, ANKRD26-like family A, member 1, CT104.3, POTE ankyrin domain family member A, POTE ankyrin domain family, member A, POTE-8ANKRD26-like family A member 1, POTE8cancer/testis antigen family 104, member 3, Prostate, ovary, testis-expressed protein on chromosome 8, protein expressed in prostate, ovary, testis, and placenta 8 |
| Host Species | Rabbit |
| Immunogen | POTE ankyrin domain family, member A. (Uniprot# Q6S8J7) |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?