missing translation for 'onlineSavingsMsg'
Learn More

Potassium tungsten oxide, 99.5% (metals basis)

Product Code. 11311569
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11311569

Brand: Thermo Scientific Alfa Aesar 014031.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-60-5 | K2O4W | 326.033 g/mol

Potassium tungsten oxide is used in solar energy and water treatment applications. It is used as alkaloids precipitating agent, bronze production and in tungsten refining. It is also used as an analytical reagent. Further, it is added to molten carbonate fuel cell to prevent the corrosion of stainless-steel separator and retards the sintering of the anode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-60-5
Molecular Formula K2O4W
Molecular Weight (g/mol) 326.033
MDL Number MFCD00049668
InChI Key AAQNGTNRWPXMPB-UHFFFAOYSA-N
Synonym potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w
PubChem CID 3084032
IUPAC Name dipotassium;dioxido(dioxo)tungsten
SMILES [O-][W](=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 921°C
Physical Form Powder
Quantity 25 g
Merck Index 14,7694
Solubility Information Soluble in water.
Formula Weight 326.04
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic
Density 3.12 g/mL
Assay (metals basis)
Chemical Name or Material Potassium tungsten oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.