missing translation for 'onlineSavingsMsg'
Learn More

Potassium tungstate, 94%, -100 mesh, Thermo Scientific™

Product Code. 12337183 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 12337183

Brand: Thermo Scientific Acros 446871000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7790-60-5
Molecular Formula K2O4W
Molecular Weight (g/mol) 326.05
InChI Key AAQNGTNRWPXMPB-UHFFFAOYSA-N
Synonym potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w
PubChem CID 3084032
IUPAC Name dipotassium;dioxido(dioxo)tungsten
SMILES [O-][W](=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 921°C
Quantity 100 g
Assay Percent Range 53 to 56% (W)
Linear Formula K2WO4
Merck Index 15,7811
Formula Weight 326.05
Percent Purity 94%
Packaging Glass Bottle
Chemical Name or Material Potassium tungstate
Product Identifier
  • Potassium tungstate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.