missing translation for 'onlineSavingsMsg'
Learn More

Potassium trioxalatoferrate(III) trihydrate

Product Code. p-7049912
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 15481177

Brand: Thermo Scientific Alfa Aesar 031124.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5936-11-8 | C6H3FeK3O12 | 440.22 g/mol

Potassium trioxalatoferrate(III) trihydrate is used in chemical actinometry to measure light flux.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5936-11-8
Molecular Formula C6H3FeK3O12
Molecular Weight (g/mol) 440.22
MDL Number MFCD00150450
InChI Key YXTWCYYDGRBUDE-UHFFFAOYSA-K
Synonym Potassium iron(III) oxalate
PubChem CID 131675837

Specifications

Color Green
Physical Form Crystalline
Quantity 5 g
UN Number UN3288
Solubility Information Slightly soluble in water.
SMILES C(=O)(C(=O)O)O.C(=O)(C(=O)[O-])O.C(=O)(C(=O)[O-])[O-].[K+].[K+].[K+].[Fe]
IUPAC Name tripotassium;2-hydroxy-2-oxoacetate;iron;oxalate;oxalic acid
Formula Weight 491.26 (437.21 Anhydrous)
Sensitivity Hygroscopic
Density 2.133 g/mL
Chemical Name or Material Potassium trioxalatoferrate(III) trihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium trioxalatoferrate(III) trihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Potassium trioxalatoferrate(III) trihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.