missing translation for 'onlineSavingsMsg'
Learn More

Potassium Trimethylsilanolate, TRC

Product Code. 30418397
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418397 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418397 Supplier Toronto Research Chemicals Supplier No. TRCP69858025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Trimethylsilanolate
CAS 10519-96-7
Molecular Formula C3H9KOSi
Quantity 25 g
Synonym 1,1,1-Trimethylsilanol Potassium Salt, (Trimethylsiloxy)potassium, Trimethylsilanol Potassium Deriv., Trimethylsilanol Potassium Salt, Potassium Trimethylsilanolate, Potassium Trimethylsiloxide, Potassium Trimethylsilyloxide, Trimethylsiloxypotassium
InChI Formula InChI=1S/C3H9OSi.K/c1-5(2,3)4;/h1-3H3;/q-1;+1
SMILES [K+].C[Si](C)(C)[O-]
IUPAC Name potassium;trimethyl(oxido)silane
Molecular Weight (g/mol) 128.29
Formula Weight 128.006
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.