missing translation for 'onlineSavingsMsg'
Learn More

Potassium trimethylsilanolate, 2M solution in THF, AcroSeal™

Product Code. 13458688 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13458688 100 mL AcroSeal™ Glass bottle 100mL
13448688 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13458688 Supplier Thermo Scientific Supplier No. 448461000
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10519-96-7 | C3H10KOSi | 129.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10519-96-7 , 109-99-9
Molecular Formula C3H10KOSi
Molecular Weight (g/mol) 129.30
MDL Number MFCD00013421
InChI Key COTHYYYVPUZALV-UHFFFAOYSA-N
Synonym potassium trimethylsilanolate, trimethylsiloxypotassium, silanol, trimethyl-, potassium salt, trimethylsilanol potassium salt, potassium trimethyl silanolate, silanol, 1,1,1-trimethyl-, potassium salt 1:1, kosime3, kotms, tmsok, c3h9osi.k
PubChem CID 23668039
SMILES [K].C[Si](C)(C)O

Specifications

Quantity 100 mL
Linear Formula (CH3)3SiOK
Flash Point −17°C
IUPAC Name potassium;trimethyl(oxido)silane
Formula Weight 128.29
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.91
Density 0.9100g/mL
Chemical Name or Material Potassium trimethylsilanolate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium trimethylsilanolate, 2M sol in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.