missing translation for 'onlineSavingsMsg'
Learn More

Potassium Trimethylacetate, TRC

Product Code. 30414751
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414751 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414751 Supplier Toronto Research Chemicals Supplier No. TRCP993515500MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Potassium Trimethylacetate
CAS 19455-23-3
Molecular Formula C5 H9 O2 . K
Quantity 500 mg
Synonym Propanoic acid','2,2-dimethyl-','potassium salt (1:1) (ACI), Pivalic acid','potassium salt (8CI), Propanoic acid','2,2-dimethyl-','potassium salt (9CI), 2,2-Dimethylpropanoic acid potassium salt, Potassium 2,2-dimethylpropanoate, Potassium pivalate, Potassium tert-butylcarboxylate, Potassium trimethylacetate, 2,2-Dimethylpropionic acid potassium salt
InChI Formula InChI=1S/C5H10O2.K/c1-5(2,3)4(6)7;/h1-3H3,(H,6,7);/q;+1/p-1
SMILES [K+].CC(C)(C)C(=O)[O-]
IUPAC Name potassium;2,2-dimethylpropanoate
Molecular Weight (g/mol) 140.222
Formula Weight 140.024
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.