missing translation for 'onlineSavingsMsg'
Learn More

Potassium trimethylacetate, 95%

Product Code. 15439278
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439278 100 g 100g
15429278 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439278 Supplier Thermo Scientific Alfa Aesar Supplier No. H62803.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19455-23-3 | C5H9KO2 | 140.223 g/mol

Potassium trimethylacetate is used as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium trimethylacetate is used as a intermediate for chemical research.

Notes
Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19455-23-3
Molecular Formula C5H9KO2
Molecular Weight (g/mol) 140.223
MDL Number MFCD00671345
InChI Key WFMNHCSATCWAAQ-UHFFFAOYSA-M
Synonym potassium pivalate, potassium trimethylacetate, pivalic acid potassium salt, potassium 2,2-dimethylpropionate, trimethylacetic acid potassium salt, 2,2-dimethylpropionic acid potassium salt
PubChem CID 23662159
IUPAC Name potassium;2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)[O-].[K+]

Specifications

Melting Point ∼200°C
Linear Formula (CH3)3CCO2K
Quantity 100 g
Formula Weight 140.22
Percent Purity 95%
Chemical Name or Material Potassium trimethylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.