missing translation for 'onlineSavingsMsg'
Learn More

Potassium tricyanomethanide, Thermo Scientific Chemicals

Product Code. 11327967
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Potassium tricyanomethanide is a precursor used for the preparation of dicyano ketene aminals. It reacts with amines to prepare alkyl ammonium tricyanomethanide. Further, it is involved in the preparation of tricyanomethanide-bridged binuclear organometallic complexes such as Cp(dppe)FeC(CN)3 and Cp(PPh3)2RuC(CN)3. It is used as starting material utilized in the preparation of 5-Amino-3-bromo-1-(tert-butyl)-1H-pyrazole-4-carbonitrile, which undergoes sandmeyer reaction to get the corresponding diaminopyrazole, 5-amino-3-aryl-1-(tert-butyl)-1H-pyrazole-4-carboxamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34171-69-2
Molecular Formula C4KN3
Molecular Weight (g/mol) 129.163
MDL Number MFCD00058850
InChI Key ZKJPYQKGNUBNOA-UHFFFAOYSA-N
Synonym potassium tricyanomethanide, potassium methanetricarbonitrile, potassium cyanoform, acmc-1ajhe, potassium ion tricyanomethanide, tricyanomethanide potassium salt, potassiomethanetricarbonitrile
PubChem CID 10964525
IUPAC Name potassium;methanetricarbonitrile
SMILES C(#N)[C-](C#N)C#N.[K+]

Specifications

Color White
Linear Formula KC(CN)3
Quantity 1 g
UN Number UN3439
Solubility Information Soluble in water.
Formula Weight 129.16
Physical Form Crystalline
Chemical Name or Material Potassium tricyanomethanide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'