missing translation for 'onlineSavingsMsg'
Learn More

Potassium trichloro(ethylene)platinate(II) monohydrate, Pt 50.5% min

Product Code. 15416585
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15416585

Brand: Thermo Scientific Alfa Aesar 044028.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16405-35-9 | C2H4Cl3KPt·H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 220°C (decomposition)
CAS 16405-35-9
Color Yellow
Physical Form Solid
Quantity 1 g
Molecular Formula C2H4Cl3KPt·H2O
Linear Formula K[PtCl3(C2H4)]·H2O
MDL Number MFCD00150524
Formula Weight 386.61 (368.59 Anhydrous)
Concentration or Composition (by Analyte or Components) Pt 50.5% min
Sensitivity Hygroscopic
Chemical Name or Material Potassium trichloro (ethylene)platinate (II) monohydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.