missing translation for 'onlineSavingsMsg'
Learn More

Potassium trans-beta-styryltrifluoroborate, 98%

Product Code. 11384855
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11384855

Brand: Thermo Scientific Alfa Aesar L17971.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 201852-49-5 | C8H7BF3K | 210.05 g/mol

Potassium trans-beta-styryltrifluoroborate is used as a reactant for Suzuki-Miyaura cross-coupling, in enantioselective total synthesis of naseseazines A and B via regioselective Friedel-Crafts based arylative dimerization, in metal-free chlorodeboronation reactions, in preparation of aryl ketones via Lewis acid-catalyzed nucleophilic substitution and in preparation of Weinreb amides by palladium-catalyzed cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium trans-beta-styryltrifluoroborate is used as a reactant for Suzuki-Miyaura cross-coupling, in enantioselective total synthesis of naseseazines A and B via regioselective Friedel-Crafts based arylative dimerization, in metal-free chlorodeboronation reactions, in preparation of aryl ketones via Lewis acid-catalyzed nucleophilic substitution and in preparation of Weinreb amides by palladium-catalyzed cross-coupling reactions.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201852-49-5
Molecular Formula C8H7BF3K
Molecular Weight (g/mol) 210.05
MDL Number MFCD02093981
InChI Key NONAUTDEFRJJII-UHFFFAOYSA-N
Synonym potassium trans-styryltrifluoroborate, potassium beta-styryltrifluoroborate, beta-styryltrifluoroboric acid potassium salt, pubchem11312, potassiostyryltrifluoroboron v, potassium-styryltrifluoroborate, potassium a-styryltrifluoroborate, potassium b-styryltrifluoroborate, potassium e-styryltrifluoroborate, potassium 2-phenylethenyltrifluoroborate
PubChem CID 23664278
IUPAC Name potassium;trifluoro-[(E)-2-phenylethenyl]boranuide
SMILES [K+].F[B-](F)(F)C=CC1=CC=CC=C1

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 7782992
Solubility Information Soluble in water.
Formula Weight 210.05
Percent Purity 98%
Chemical Name or Material Potassium trans-beta-styryltrifluoroborate
compliance-icons
Product Identifier
  • Potassium trans-beta-styryltrifluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Potassium trans-styryltrifluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.