missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiosulfate hydrate, 96+%, extra pure

Product Code. 10035213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10035213 100 g Plastic bottle 100g
10638044 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10035213 Supplier Thermo Scientific Acros Supplier No. 388691000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1264198-56-2 | K2O3S2 | 190.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1264198-56-2
Molecular Formula K2O3S2
Molecular Weight (g/mol) 190.31
MDL Number MFCD00150405
InChI Key FGRVOLIFQGXPCT-UHFFFAOYSA-L
PubChem CID 61501
SMILES [K+].[K+].[O-]S([S-])(=O)=O

Specifications

Color Colorless to Yellow
pH 6 to 9 (5% soln.)
Physical Form Liquid
Quantity 100 g
Assay Percent Range 96.0 to 104% (On dry substance) (Iodimetry)
Linear Formula K2S2O3·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
IUPAC Name dipotassium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Formula Weight 190.33
Percent Purity ≥96%
Grade Extra Pure
Loss on Drying 4% max. (150°C)
Packaging Plastic bottle
Chemical Name or Material Potassium thiosulfate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.