missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, ACS Reagent, ≥99%, Solstice

Product Code. 16605803
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16605803 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16605803 Supplier Fluka Supplier No. 3127225KG
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured

TRUSTED_SUSTAINABILITY

Specifications

CAS 333-20-0
Color Light Yellow
Quantity 25 kg
Molecular Formula CKNS
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
SMILES [K+].[S-]C#N
IUPAC Name potassium cyanosulfanide
Molecular Weight (g/mol) 97.18
Chemical Name or Material Potassium thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.