missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, ACS, 98.5% min

Product Code. 11327887
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327887 500 g 500g
11337887 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327887 Supplier Thermo Scientific Alfa Aesar Supplier No. 014318.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 333-20-0 | CKNS | 97.176 g/mol

Potassium thiocyanate is used in the preparation of ethylenesulfide, lead thiocyante, which is used in the convertion of acyl chloride to thiocyanates. It is also used in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury-thiocyanate and selenocyanate compounds as potential models. It finds application in cosmetic and personal care product formulations as a skin conditioning agent. Also used in moisturizers and oral care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Boiling Point 500°C (decomposition)
Melting Point 172°C to 175°C
Color White
Physical Form Crystals
Quantity 500 g
Assay Percent Range 98.5% min.
Beilstein 3594799
Solubility Information Soluble in water,acetone,and alcohol. When dissolved in its own weight of water,the temperature drops ^=30° Soluble in water,acetone and alcohol.
Formula Weight 97.18
Grade ACS
Odor Odorless
Sensitivity Light and moisture sensitive
Appearance White crystalline
Density 1.89
Chemical Name or Material Potassium thiocyanate, For ACS analysis
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.