missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, 98%, pure

Product Code. 10736753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10736753 2.5 kg Plastic bottle 2.5kg
10349232 100 g Plastic bottle 100g
10461533 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10736753 Supplier Thermo Scientific Acros Supplier No. 377430025
  • ONLINE EXCLUSIVE PRICE
    £282.00
    £273.00 / 2.5kg
    Save £9.00
    3% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 333-20-0 | CKNS | 97.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.18
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]

Specifications

Melting Point 170.0°C to 179.0°C
Color Colorless or White
Physical Form Crystals
Quantity 2.5 kg
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula KSCN
Merck Index 15,7807
Solubility Information Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 97.18
Percent Purity 98%
Grade Pure
Packaging Plastic bottle
Specific Gravity 1.886
Density 1.8860g/mL
Chemical Name or Material Potassium thiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.