missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Potassium tetraphenylborate, 97%

Brand:  Alfa Aesar™ H55276.14

Code : NEW

Additional Details : CAS Number : 3244-41-5 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15471478

  • £149.10 / 25g


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Description

Description

Applications
It is an active pharmaceutical intemediate.

Solubility
Low solubility in water. Soluble in organic solvents.

Notes
Desiccate at room temperature. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

Potassium tetraphenylborate
KB(C6H5)4
MFCD03093877
3244-41-5
358.33

Specifications

Potassium tetraphenylborate
Powder
KB(C6H5)4
MFCD03093877
>300°C
3244-41-5
97%
358.33
White
25g
Videos
SDS