missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tetraoxalate Dihydrate, ≥97.0% (RT), Honeywell™ Fluka™

Product Code. 15695990 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15695990 1 kg Poly bottle 1kg
15606000 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15695990 Supplier Honeywell-Fluka Supplier No. 605941KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6100-20-5
Molecular Formula C4H3KO8
Molecular Weight (g/mol) 218.16
MDL Number MFCD00150443,MFCD00150443
InChI Key GANDVAJEIJXBQJ-UHFFFAOYSA-M
Synonym potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate
PubChem CID 131698588
IUPAC Name potassium oxalic acid hydrogen oxalate
SMILES [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O

Specifications

Melting Point >160°C
Color Colorless
pH 1.7 (at 12.60g/L, 25°C)
Physical Form Solid
Quantity 1 kg
Linear Formula KH3(C2O4)2 · 2H2O
UN Number NONH for all modes of transport
Beilstein 3854419
Formula Weight 254.19g/mol
Percent Purity ≥97.0% (RT)
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Potassium tetraoxalate dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.